Skip to Content
Merck
All Photos(3)

Key Documents

T6541

Sigma-Aldrich

TES

BioXtra, ≥99.5% (titration)

Synonym(s):

2-[(2-Hydroxy-1,1-bis(hydroxymethyl)ethyl)amino]ethanesulfonic acid, N-[Tris(hydroxymethyl)methyl]-2-aminoethanesulfonic acid, TES free acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15NO6S
CAS Number:
Molecular Weight:
229.25
Beilstein:
1957061
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Assay

≥99.5% (titration)

form

powder

impurities

Insoluble matter, passes filter test

ign. residue (900 °C)

≤0.1% (as SO4)

loss

≤1% loss on drying, 110°C

pH

3.5-5.0 (20 °C, 1 M in H2O)

useful pH range

6.8-8.2

pKa (25 °C)

7.5

mp

~225 °C (dec.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%
As: ≤0.0001%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.001%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.02%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
Na: ≤0.01%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

absorption

≤0.045 at 280 in H2O at 1 M
≤0.055 at 260 in H2O at 1 M

SMILES string

OCC(CO)(CO)NCCS(O)(=O)=O

InChI

1S/C6H15NO6S/c8-3-6(4-9,5-10)7-1-2-14(11,12)13/h7-10H,1-5H2,(H,11,12,13)

InChI key

JOCBASBOOFNAJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Sugiyama et al.
Molecular reproduction and development, 58(1), 30-38 (2001-01-06)
The growth, differentiation, and death/survival of spermatogonia are precisely regulated for the proper production of spermatozoa. We have previously shown that Bcl-2 ectopically expressed in spermatogonia caused the inhibition of normal spermatogonial apoptosis and the subsequent failure of differentiation in
R H Martin et al.
Biology of reproduction, 47(2), 268-270 (1992-08-01)
Human sperm karyotypes can be prepared after fusion of human sperm with Golden hamster oocytes. Most laboratories use one of two methods of sperm capacitation: incubation of freshly-ejaculated sperm in Biggers, Whitten, and Whittingham (BWW) medium for 5-7 h at
Yikang Wu et al.
Organic letters, 4(13), 2141-2144 (2002-06-21)
[reaction: see text] In DMSO cleavage of triethylsilyl (TES) ethers by o-iodoxybenzoic acid (IBX) was significantly faster than cleavage of tert-butyldimethylsilyl (TBS) ethers or further oxidation into carbonyl compounds. In most cases, TES protecting groups could be removed in good
M J Taylor et al.
The British journal of ophthalmology, 73(10), 781-791 (1989-10-01)
Preservation solutions for short-term storage of isolated donor corneas for use in penetrating keratoplasty have all been based on tissue culture medium, on the assumption that media designed to maintain the viability of cells at physiological temperatures will also provide
Hao Li et al.
Organic letters, 9(6), 965-968 (2007-02-15)
A conjugate addition-aldol-dehydration reaction of alpha,beta-unsaturated aldehydes with 2-N-protected amino benzaldehydes has been developed. The process is promoted by (S)-diphenylprolinol TES ether to afford synthetically useful 1,2-dihydroquinolines in high enantioselectivities with good yields. [reaction: see text]

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service