Skip to Content
Merck
All Photos(1)

Key Documents

T6041

Supelco

Testosterone decanoate

≥98% (TLC)

Synonym(s):

17β-Hydroxy-4-androsten-3-one 17-decanoate, 4-Androsten-17β-ol-3-one 17-caprate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H46O3
CAS Number:
Molecular Weight:
442.67
EC Number:
MDL number:
UNSPSC Code:
41116107

Assay

≥98% (TLC)

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

SMILES string

CCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C29H46O3/c1-4-5-6-7-8-9-10-11-27(31)32-26-15-14-24-23-13-12-21-20-22(30)16-18-28(21,2)25(23)17-19-29(24,26)3/h20,23-26H,4-19H2,1-3H3/t23-,24-,25-,26-,28-,29-/m0/s1

InChI key

LBERVHLCXUMDOT-MPZZESAYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Haidl et al.
Der Urologe. Ausg. A, 47(12), 1555-1556 (2008-11-18)
Evidence-based drug therapy for male infertility is often difficult because 30% of all cases of male infertility are classified as idiopathic, and another 30% need surgical treatment. Without knowledge of the underlying pathology, there is no foundation for a specific
B M Brady et al.
Human reproduction (Oxford, England), 21(1), 285-294 (2005-09-21)
The combination of etonogestrel implants with injectable testosterone decanoate was investigated as a potential male contraceptive. One hundred and thirty subjects were randomly assigned to three treatment groups, all receiving two etonogestrel rods (204 mg etonogestrel) and 400 mg testosterone
Cathy J Hay et al.
The Journal of clinical endocrinology and metabolism, 90(4), 2042-2049 (2005-01-27)
The effect of a novel combination of oral etonogestrel (ENG) and im testosterone decanoate (TD) on suppression of gonadotropins and spermatogenesis as a potential lead for male contraception was investigated. Healthy male volunteers were randomized into two groups receiving 300
Determination of testosterone, testosterone propionate, testosterone enanthate, testosterone undecanoate in pharmaceutical preparations by HPLC.
S Claussen et al.
Die Pharmazie, 55(2), 147-148 (2000-03-21)
John K Amory
Current opinion in endocrinology, diabetes, and obesity, 15(3), 255-260 (2008-04-29)
Testosterone functions as a contraceptive by suppressing the secretion of luteinizing hormone and follicle-stimulating hormone from the pituitary. Low concentrations of these hormones deprive the testes of the signals required for spermatogenesis and results in markedly decreased sperm concentrations and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service