Skip to Content
Merck
All Photos(1)

Key Documents

T5700

Sigma-Aldrich

Anti-Thrombin Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

AI482343, ALPHA THROMBIN RECEPTOR, CF2R, F2R, HTR, MGC28086, MGC93622, PAR-1, THROMBIN RECEPTOR, TRGPC, ThR, ThrR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 7 μg/mL using human bone marrow megakaryocytes

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... F2R(2149)
mouse ... F2r(14062)
rat ... F2r(25439)

Immunogen

synthetic peptide corresponding to the N-terminal extracellular domain of human thrombin receptor, conjugated to KLH. The immunizing peptide has a mouse homology of 42% (Identity) and 58% (Similarity).

Target description

Thrombin Receptor, coagulation factor II receptor, is a 7-transmembrane receptor involved in the regulation of thrombotic response.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.1% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service