Skip to Content
Merck
All Photos(1)

Key Documents

T3384

Sigma-Aldrich

L-Tyrosine β-naphthylamide

≥98% (TLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€438.00

€438.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€438.00

About This Item

Empirical Formula (Hill Notation):
C19H18N2O2
CAS Number:
Molecular Weight:
306.36
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

€438.00


Check Cart for Availability

Request a Bulk Order

Product Name

L-Tyrosine β-naphthylamide,

Assay

≥98% (TLC)

Quality Level

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

N[C@@H](Cc1ccc(O)cc1)C(=O)Nc2ccc3ccccc3c2

InChI

1S/C19H18N2O2/c20-18(11-13-5-9-17(22)10-6-13)19(23)21-16-8-7-14-3-1-2-4-15(14)12-16/h1-10,12,18,22H,11,20H2,(H,21,23)/t18-/m0/s1

InChI key

BZBYYTSHGRKCJJ-SFHVURJKSA-N

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M de Gandarias et al.
Arzneimittel-Forschung, 49(10), 816-819 (1999-11-11)
Lithium (CAS 7439-93-2) is being utilized widely for the treatment of manic-depressive illness. However, the neurochemical mechanism of the ion action still remains relatively obscure. In the present work, the effect of acute lithium administration on soluble and membrane-bound enkephalin-degrading
J M de Gandarias et al.
Life sciences, 59(13), 1097-1101 (1996-01-01)
Puromycin sensitive and insensitive membrane-bound aminopeptidase activity levels during the estrous cycle in several brain areas have been described in this research. We have found the highest aminopeptidase M activity levels during the proetrous stage in the hypothalamus, the amygdala
J Negrel et al.
Phytochemistry, 56(6), 523-527 (2001-04-03)
L-Tyrosine beta-naphthylamide, a synthetic substrate designed to measure tyrosine aminopeptidase activity, is a potent inhibitor of hydroxycinnamoyl-CoA:tyramine N-(hydroxycinnamoyl)transferase (THT) purified from elicited tobacco cell-suspension cultures. The inhibition is competitive, with the inhibitor binding reversibly to the tyramine binding site of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service