Skip to Content
Merck
All Photos(1)

Key Documents

T2700

SAFC

TWEEN® 20

meets EP testing specifications

Synonym(s):

Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.25
Pricing and availability is not currently available.

Agency

meets EP testing specifications

Quality Level

description

non-ionic

form

liquid

mol wt

~1228

composition

lauric acid, ≥40% (balance primarily myristic, palmoyic, and stearic acids)

refractive index

n20/D 1.468 (lit.)

CMC

0.059 mM

transition temp

cloud point 76 °C

density

1.095 g/mL at 25 °C (lit.)

HLB

16.7

SMILES string

CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TWEEN® 20, also known as polysorbate 20, is a non-ionic detergent. It reduces the surface tension in solutions and thereby aids in uniform spreading of reagents. Tween 20 has a gentle action on cell lipids, preserves the integrity of cell surface antigens and aids in the staining of intracellular proteins.[1][2]

Application

Non-ionic detergent
TWEEN 20 has been used as a surfactant and has also been used for enzyme extraction.[3][4]

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

527.0 °F - Pensky-Martens closed cup

Flash Point(C)

275 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production and extraction of an extracellular lipase from the entomopathogenic fungus Metarhizium anisopliae
Walter Orlando Beys Silva
Process. Biochem. (2005)
PAHs soil decontamination in two steps: Desorption and electrochemical treatment
M. Teresa Alcantara
Journal of Hazardous Materials (2009)
Permeability of Biological Membranes (2016)
Immunohistochemistry: Basics and Methods (2010)
Aida Maddahi et al.
BMC neuroscience, 9, 85-85 (2008-09-17)
MEK1/2 is a serine/threonine protein that phosphorylates extracellular signal-regulated kinase (ERK1/2). Cerebral ischemia results in enhanced expression of cerebrovascular contractile receptors in the middle cerebral artery (MCA) leading to the ischemic region. Here we explored the role of the MEK/ERK

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service