Skip to Content
Merck
All Photos(1)

Key Documents

T2069

Sigma-Aldrich

Trizma® hydrochloride solution

pH 7.2, BioPerformance Certified, 1 M, suitable for cell culture

Synonym(s):

Tris hydrochloride solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€59.60
1 L
€168.00

€59.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€59.60
1 L
€168.00

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

€59.60


Check Cart for Availability

Request a Bulk Order

grade

BioPerformance Certified
Molecular Biology
for molecular biology

form

solution

concentration

1 M

technique(s)

cell culture | mammalian: suitable

impurities

DNase, RNase, Protease, none detected
bioburden, tested
endotoxin, tested
≤5 ppm Heavy metals (as Pb)

pH

7.2

useful pH range

7.0-9.0

absorption

≤0.05 at 290 at 40%

Looking for similar products? Visit Product Comparison Guide

General description

A series of Pre-mixed solutions of TRIZMA Base and TRIZMA HCl to provide commonly used pH values for Tris buffers. No mixing or pH adjustment necessary. Guaranteed accuracy ± 0.1 pH units.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service