Skip to Content
Merck
All Photos(1)

Documents

T1788

Sigma-Aldrich

Tyrode′s Solution, Acidic

liquid, sterile-filtered, suitable for mouse embryo cell culture

Synonym(s):

Cell culture solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | embryo: suitable

impurities

endotoxin, tested

pH

2.5±0.3

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Tyrode′s salt solution is used for diluting protein components of media of natural origin. The current role of a balanced salt solution in cell culture is multifaceted. It serves as an irrigating, transporting, and diluting agent while maintaining intra- and extracellular osmotic balance and provides cells with water and certain bulk inorganic ions essential for normal cell metabolism. It also provides the essential energy for cell metabolism and a buffering system to maintain the medium within the optimal pH range (7.2-7.6).

Application

Tyrode′s Solution, Acidic has been used for the removal of zona pellucida from oocytes/embryos.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Carvalhal et al.
Molecular biology of the cell, 28(19), 2470-2478 (2017-08-05)
Asymmetric cell divisions depend on the precise placement of the spindle apparatus. In mammalian oocytes, spindles assemble close to the cell's center, but chromosome segregation takes place at the cell periphery where half of the chromosomes are expelled into small
Zhao-Yang Xu et al.
Scientific reports, 5, 16978-16978 (2015-11-20)
Cep55 is a relatively novel member of the centrosomal protein family. Here, we show that Cep55 is expressed in mouse oocytes from the germinal vesicle (GV) to metaphase II (MII) stages. Immuostaining and confocal microscopy as well as time lapse
Liyan Sui et al.
Journal of cellular physiology, 235(10), 7030-7042 (2020-02-06)
Histone deacetylase 6 (HDAC6) participates in mouse oocyte maturation by deacetylating α-tubulin. However, how HDAC6 expression is regulated in oocytes remains unknown. In the present study, we discovered that mouse oocytes had a high level of HDAC6 expression and a
Jiangwei Lin et al.
Scientific reports, 6, 39457-39457 (2016-12-20)
Various types of stem cell lines have been derived from preimplantation or postimplantation mouse embryos: embryonic stem cell lines, epiblast stem cell lines, and trophoblast stem cell lines. It is not known if extraembryonic endoderm stem (XEN) cell lines can
Qianhua Xu et al.
Nature genetics, 51(5), 844-856 (2019-05-02)
The oocyte epigenome plays critical roles in mammalian gametogenesis and embryogenesis. Yet, how it is established remains elusive. Here, we report that histone-lysine N-methyltransferase SETD2, an H3K36me3 methyltransferase, is a crucial regulator of the mouse oocyte epigenome. Deficiency in Setd2

Articles

Mouse embryo media and embryo validated reagents for transgenic mouse embryo culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service