Skip to Content
Merck

Skip To

T0950

Anti-γ-Tubulin (QG-17) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
12


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

Drosophila

technique(s)

western blot: 0.2-0.4 μg/mL using whole cell extract of the Schneider′s Drosophila Line 2 [D.mel.(2), SL2]

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

γ-Tubulin (48 kDa) is a widely expressed and highly conserved protein within the microtubule organizing centers (MTOCs) or centrosome in eukaryotic cells.
γ-tubulin, mapped on human chromosome 17q21.2, codes for a member of the tubulin family. It is localized to the microtubule organizing centres. γ-tubulin consists of two isoforms tubulin γ 1 (TUBG1) and tubulin γ 2 (TUBG2) with 97.3% amino acid identity. In addition to these two isoforms, γ-tubulin pseudogene (TUBG1P) is also been identified on human chromosome 17. Human TUBG1 and TUBG2 transcripts are widely expressed in preimplantation embryos and the brain, respectively. γ-tubulin is a component of γ-tubulin ring complex (γ-TuRC), which has roughly the same diameter as a microtubule.

Immunogen

synthetic peptide corresponding to the C-terminus of Drosophila γ-tubulin (amino acids 441-457 with N-terminal added cysteine-glycine), conjugated to KLH.

Application

Anti-γ-Tubulin (QG-17) antibody has been used in immunoblotting and immunostaining.

Biochem/physiol Actions

γ-tubulin nucleates microtubule assembly throughout the mammalian cell cycle in vivo. In Aspergillus nidulans, γ-tubulin facilitates attachment of microtubules to the spindle pole body, nuclear division and microtubule assembly. Ubiquitination of γ-tubulin by breast cancer 1 protein (BRCA1) is a crucial step in the regulation of centrosome number. Overexpression of γ-tubulin is observed in lung cancer. Tubulin γ 2 (TUBG2) plays a vital role in cell growth. Aberration in the γ-tubulin gene alters microtubule assembly. The expression levels of γ-tubulin can be considered as an important prognostic indicator for patients with astrocytomas.
The sequence is highly specific for Drosophila γ-tubulin and is not found in γ-tubulin of other species or other members of the tubulin family such as α-, β-, δ-, and ε-tubulins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T3195T1450T5326
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

antibody form

purified antibody (purified from hybridoma cell culture)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

GTU-88, monoclonal

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters