Skip to Content
Merck
All Photos(1)

Key Documents

T0202

Sigma-Aldrich

Tocainide hydrochloride

≥98% (HPLC), solid

Synonym(s):

2-Amino-N-(2,6-dimethylphenyl)propanamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€186.00
50 MG
€737.00

€186.00


Estimated to ship on17 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€186.00
50 MG
€737.00

About This Item

Empirical Formula (Hill Notation):
C11H16N2O · HCl
CAS Number:
Molecular Weight:
228.72
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€186.00


Estimated to ship on17 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated
under inert gas

color

white

solubility

DMSO: >20 mg/mL
H2O: ≥5 mg/mL

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

Cl[H].CC(N)C(=O)Nc1c(C)cccc1C

InChI

1S/C11H16N2O.ClH/c1-7-5-4-6-8(2)10(7)13-11(14)9(3)12;/h4-6,9H,12H2,1-3H3,(H,13,14);1H

InChI key

AMZACPWEJDQXGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tocainide hydrochloride may be used in cell signaling studies.

Biochem/physiol Actions

Tocainide hydrochloride is a primary amine analog of lidocaine (lignocaine), used for the treatment of tinnitus.[1] It blocks the sodium channels in the pain-producing foci in the nerve membranes and renders an analgesic effect in trigeminal neuralgia.[2]
Tocainide hydrochloride is a sodium channel blocker; Class IB antiarrhythmic.

Features and Benefits

This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.
Tocainide is voltage-dependent and use-dependent sodium channel blocker (class IB type antiarrhythmic); Ki = 115 μM in blockade of inactivated-state sodium channels; IC50 = 985μM in blockade of Na+ currents at a holding potential (HP) of -140 mV; IC50 = 254 μM in blockade of Na+ currents at HP -70 mV; IC50 = 523 μM in blockade of Na+ currents at tonic block (-100 mV); IC50 = 248 μM for use-dependent block at 10 Hz.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Carrieri et al.
European journal of medicinal chemistry, 44(4), 1477-1485 (2008-11-26)
Enantiomeric forms of Tocainide, Mexiletine, and structurally related local anaesthetic compounds, were analyzed with respect to their potency in blocking Na(v)1.4 channel. Structure-activity relationships based on in vitro pharmacological assays, suggested that an increase in terms of lipophilicity and/or molecular
L He et al.
The Cochrane database of systematic reviews, (3)(3), CD004029-CD004029 (2006-07-21)
Non-antiepileptic drugs have been used in trigeminal neuralgia management since the 1970s. The objective was to review systematically the efficacy of non-antiepileptic drugs for trigeminal neuralgia. We searched the Cochrane Neuromuscular Disease Group Register, MEDLINE, EMBASE, and LILACS (all to
Annamaria De Luca et al.
Neuromuscular disorders : NMD, 22(1), 56-65 (2011-08-02)
Drug screening on sodium currents of native myofibers by means of voltage-clamp recordings is predictive of pre-clinical anti-myotonic activity in vivo and ex vivo. By this approach we identified the N-benzylated beta-proline derivative of tocainide (To10) as the most potent
C Ghelardini et al.
Neuroscience, 169(2), 863-873 (2010-06-29)
The role of voltage-gated sodium channels in the transmission of neuropathic pain is well recognized. For instance, genetic evidence recently indicate that the human Nav1.7 sodium channel subtype plays a crucial role in the ability to perceive pain sensation and
Alessia Catalano et al.
European journal of medicinal chemistry, 43(11), 2535-2540 (2008-03-18)
1-Benzyl-N-(2,6-dimethylphenyl)piperidine-3-carboxamide and 4-benzyl-N-(2,6-dimethylphenyl)piperazine-2-carboxamide, two conformationally restricted analogues of tocainide, were designed and synthesized as voltage-gated skeletal muscle sodium channel blockers. They showed, with respect to tocainide, a marked increase in both potency and use-dependent block.

Articles

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service