SYNS389200
Synthware™ long syringe needle
O.D. × L 0.9 mm × 200 mm , Bevel Tip, needle, female luer lock
Synonym(s):
Synthware S389200, bevel tip needle, stainless steel syringe needle
About This Item
feature
needle female luer lock
packaging
pack of 1 ea
manufacturer/tradename
Kemtech S389200
O.D. × L
0.9 mm × 200 mm , Bevel Tip
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service