SYNF264525
Synthware™ Burets
class A, clear borosilicate glass 3.3, volume 25 mL
Synonym(s):
Synthware F264525, burette
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear borosilicate glass 3.3
feature
class A
packaging
pack of 1 ea
manufacturer/tradename
Kemtech F264525
volume
25 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Burets, Class A, Capacity: 25mL Clear Glass
Application
kemetch; synthware; buret; class a; clear
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service