Skip to Content
Merck
All Photos(1)

Key Documents

SRP8035

Sigma-Aldrich

IL-36α mouse

>95% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized

Synonym(s):

Interleukin-1 family member 6, Interleukin-1 homolog 1, Interleukin-1α, Interleukin-36α

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€466.00

€466.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€466.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€466.00


Usually ships in 2 weeks.

Product Name

IL-36α mouse, recombinant, expressed in E. coli, untagged, >95% (SDS-PAGE)

biological source

mouse

recombinant

expressed in E. coli

conjugate

untagged

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~17 kDa

packaging

pkg of 10 μg

impurities

<0.01 EU/μg endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il36a(54448)

General description

IL-36α (IL-1F6), IL-36β (IL-1F8) and IL-36γ (IL-1F9) bind to IL-36R (IL-1Rrp2) and IL-1RAcP, activating similar intracellular signals as IL-1. IL-36Ra inhibits the production of proinflammatory cytokines, including IL-12, IL-1β, IL-6, TNF-α and IL-23 induced by IL-36 in BMDC and CD4 T cells. Skin and dendritic cells are targets of the IL-36 interleukins leading to a Th1 response. These cytokines may represent potential targets for immune-mediated inflammatory conditions or, alternatively, could be used as adjuvants in vaccination.

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute with 100 μL deionized water.

Other Notes

IL-36alpha (mouse) (aa 8-160) is untagged.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service