Skip to Content
Merck
All Photos(1)

Key Documents

SRP8021

Sigma-Aldrich

NMNAT1 human

recombinant, expressed in E. coli, His tagged, >95% (SDS-PAGE)

Synonym(s):

EC 2.7.7.1, IL-1F9, NMN adenylyltransferase 1, Nicotinamide mononucleotide adenylyltransferase 1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€263.00

€263.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€263.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

€263.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

tag

His tagged

Assay

>95% (SDS-PAGE)

form

liquid

mol wt

monomer ~33.3 kDa

packaging

pkg of 10 μg

concentration

≥0.2 mg/mL

color

clear

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... NMNAT1(64802)

General description

Nuclear NMNAT isoform. Catalyzes the formation of NAD+ from nicotinamide mononucleotide (NMN) and ATP. It can also use the deamidated form of nicotinic acid mononucleotide (NAMN) as substrate with the same efficiency. Interacts with PARP-1/ARTD1. Protects against axonal degeneration following mechanical or toxic insults. Widely expressed.

Physical form

Solution in 50 mM sodium phosphate, pH 8.0, containing 300 mM sodium chloride, 2 mM DTT and 10% glycerol.

Other Notes

Human full-length NMNAT1 (aa 1-279) is fused at the N-terminus to a His-tag.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service