Skip to Content
Merck
All Photos(1)

Documents

SRP8020

Sigma-Aldrich

NAD Kinase human

recombinant, expressed in E. coli, His tagged, >95% (SDS-PAGE)

Synonym(s):

IL-1F8, NADK, Poly(P)/ATP NAD kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

tag

His tagged

Assay

>95% (SDS-PAGE)

form

liquid

mol wt

monomer ~50 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

≥0.2 mg/mL

color

clear

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... NADK(65220)

General description

NAD kinase catalyzes the transfer of a phosphate group from ATP to NAD+ to generate NADP+, which in its reduced form acts as an electron donor for biosynthetic reactions. NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis.

Physical form

Ammonium sulphate suspension.

Other Notes

Human full length NAD kinase (aa 1-446) is fused at the N-terminus to a His-tag.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Zhang et al.
Cell metabolism, 33(1), 94-109 (2020-11-08)
The emergence of cancer from diverse normal tissues has long been rationalized to represent a common set of fundamental processes. However, these processes are not fully defined. Here, we show that forced expression of glucose-6-phosphate dehydrogenase (G6PD) affords immortalized mouse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service