Skip to Content
Merck
All Photos(1)

Key Documents

SRP8009

Sigma-Aldrich

GITRL, Soluble mouse

recombinant, expressed in HEK 293 cells, FLAG® tagged, >90% (SDS-PAGE)

Synonym(s):

AITRL, EGF-like protein 9, Glucocorticoid-induced TNF receptor ligand, TNFSF18

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€564.00

€564.00


Usually ships in 2 weeks.


Select a Size

Change View
50 μG
€564.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€564.00


Usually ships in 2 weeks.

biological source

mouse

recombinant

expressed in HEK 293 cells

tag

FLAG® tagged

Assay

>90% (SDS-PAGE)

form

liquid

mol wt

~22 kDa by SDS-PAGE

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

≥0.2 mg/mL

impurities

<0.1 EU/μg endotoxin, tested

color

clear

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

GITRL (Glucocorticoid-induced TNF receptor ligand) is expressed on dendritic cells (DC), monocytes, macrophages, B cells, activated T cells, endothelial cells, osteoclasts and various healthy non-lymphoid tissues (e.g. testis). GITRL is constitutively expressed and released as soluble form by solid tumors and various hematopoietic malignancies. GITRL causes differentiation of osteoclasts, activation of macrophages, but also alteration of carcinoma and leukemia cells and influences apoptosis. Binding to GITR is important in regulating T cell proliferation and TCR-mediated apoptosis. GITRL is implicated in development of autoimmune diseases and in the immune response against infectious pathogens and tumors.

Physical form

Solution in 10 mM glycine, pH 10.0.

Other Notes

The extracellular domain of mouse GITRL (aa 45-173) is fused at the N-terminus to a FLAG®-tag.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service