SRP6533
RBP4 from mouse
recombinant, expressed in HEK 293 cells, ≥90% (SDS-PAGE)
Synonym(s):
PRBP, Plasma retinol-binding protein, RBP, Retinol Binding Protein 4
About This Item
biological source
mouse
recombinant
expressed in HEK 293 cells
Assay
≥90% (SDS-PAGE)
form
liquid
mol wt
monomer ~230 kDa (Mouse RBP4 aa 19-201 is fused at the N-terminus to a FLAG0174-tag.)
packaging
pkg of 10 μg
pkg of 50 μg
impurities
<0.1 EU/μg endotoxin (LAL test)
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... RBP4(19662)
General description
Physical form
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service