Skip to Content
Merck
All Photos(1)

Key Documents

SRP6526

Sigma-Aldrich

FTO human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

FATSO, Fat Mass and Obesity-associated Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

liquid

mol wt

~65 kDa (Human FTO aa 2-505 is fused at the N-terminus to a His-tag.)

packaging

pkg of 10 μg

concentration

0.2 mg/mL

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FTO(79068)

General description

FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse ‘fused toes′ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.
The fat mass and obesity-associated (FTO) gene is mapped to human chromosome 16q12.2. The encoded protein is a nuclear protein and belongs to the AlkB-related non-haem iron- and 2-oxoglutarate-dependent dioxygenase family.

Biochem/physiol Actions

FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse ‘fused toes′ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower high-density lipoprotein (HDL)-cholesterol. FTO stimulates adipogenesis, tumorigenesis, and drug resistance of cancer cells. It does so by reducing the levels of messenger RNA N6-methyladenosine (m6A) of certain genes post-transcription. The FTO single-nucleotide polymorphisms (SNPs) are associated with elevated body mass and obesity in humans.

Physical form

0.2 mg/mL of 0.2 μm-filtered solution in 55 mM Tris-Cl, pH 8.2 containing 150 mM NaCl.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Lan et al.
Frontiers in genetics, 11, 559138-559138 (2020-12-12)
In recent years, the prevalence of obesity and cancer have been rising. Since this poses a serious threat to human health, the relationship between the two has attracted much attention. This study examined whether fat mass and obesity-associated (FTO) genes
Katherine A Fawcett et al.
Trends in genetics : TIG, 26(6), 266-274 (2010-04-13)
In 2007, an association of single nucleotide polymorphisms (SNPs) in the fat mass and obesity-associated (FTO) gene region with body mass index (BMI) and risk of obesity was identified in multiple populations, making FTO the first locus unequivocally associated with
Xiaolan Deng et al.
Frontiers in endocrinology, 9, 396-396 (2018-08-15)
Fat mass and obesity-associated protein (FTO) single-nucleotide polymorphisms (SNPs) have been linked to increased body mass and obesity in humans by genome-wide association studies (GWAS) since 2007. Although some recent studies suggest that the obesity-related SNPs in FTO influence obesity
Sarah Boissel et al.
American journal of human genetics, 85(1), 106-111 (2009-06-30)
FTO is a nuclear protein belonging to the AlkB-related non-haem iron- and 2-oxoglutarate-dependent dioxygenase family. Although polymorphisms within the first intron of the FTO gene have been associated with obesity, the physiological role of FTO remains unknown. Here we show
Rachel M Freathy et al.
Diabetes, 57(5), 1419-1426 (2008-03-19)
Common variation in the FTO gene is associated with BMI and type 2 diabetes. Increased BMI is associated with diabetes risk factors, including raised insulin, glucose, and triglycerides. We aimed to test whether FTO genotype is associated with variation in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service