Skip to Content
Merck
All Photos(1)

Documents

SRP6522

Sigma-Aldrich

Ubiquitin-TAMRA-labeled (TMR-Ub)

≥95% (SDS-PAGE)

Synonym(s):

CEP80, HUBCEP80, RPS27A, Ribosomal Protein S27a, UBA80, UBB, UBCEP1, UBCEP80

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

9.133 kDa

packaging

pkg of 50 μg

concentration

8 mg/mL

shipped in

dry ice

storage temp.

−70°C

General description

Post-translational modification of proteins by ubiquitin (Ub) is a key regulatory process that impacts almost all cellular functions. Ubiquitylation occurs through isopeptide linkage between the C-terminus of Ub and the e-amino group of a lysine (Lys) residue on the target substrate [1]. Ub itself has seven Lys residues (6, 11, 27, 29, 33, 48, and 63), any of which can participate in further ubiquitylation, generating polyUb chains [2, 3]. Monitoring the ubiquitylation of target proteins or the growth of polyubiquitin chains has traditionally been carried out with either radiolabeled or epitope-tagged ubiquitin requiring long and laborious detection methods. Fluorescently labeled ubiquitin provides a rapid, facile technique for studying ubiquitin conjugation in vitro. Unlike others, TheTAMRA-labeled ubiquitin carries a single TAMRA molecule attached at a defined location and avoids modification of either the N-terminus or Lys side chains.

Physical form

≥ 8 mg/mL in PBS.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service