Skip to Content
Merck
All Photos(1)

Key Documents

SRP6465

Sigma-Aldrich

Lipocalin-2/LCN2 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

24p3, MSFI, NGAL, Neutrophil Gelatinase-associated Lipocalin, Oncogene 24p3, Siderocalin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 22 kDa
observed mol wt 23-25 kDa (DTT reduced. Gln 21 is the predicted N-terminal.)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LCN2(3934)

General description

LCN2 (lipocalin 2), also known as oncogene 24p3, uterocalin, siderocalin, is a 24kDa secreted glycoprotein. It was initially identified in a mouse kidney cell culture infected with simian virus 40 (SV-40). It is a member of the transporter protein family called lipocalins which transport small, hydrophobic ligands.[1]
Lipocalin-2 (LCN2), a secreted glycoprotein, is an adipokine.[2][3] It is expressed in various cells like respiratory cells, intestinal epithelial cells, endothelial cells, renal tubular cells and cardiomyocytes.[4] The gene LCN2 is located on human chromosome 9q34.[5]

Biochem/physiol Actions

LCN2 (lipocalin 2) functions as a biomarker for various inflammatory disorders as well as cancers. It is also thought to be involved in resistance to drug-mediated apoptosis. It interacts with matrix metalloproteinase-9 and promotes invasion, and it also facilitates tumor growth and is linked with poor prognosis. In lung adenocarcinoma this protein facilitates epithelial to mesenchymal transition (EMT), and hence, promotes metastasis and tumorigenesis, which is repealed by BRM270, an NFκB inhibitor (nuclear factor).[6] Urine levels of LCN2 increase post-nephrotoxic and ischemic insult. In patients undergoing off pump coronary bypass surgery (OPCAB), this protein functions as an early biomarker for acute kidney injury.[7]
Lipocalin-2 (LCN2) has been implicated in a variety of processes including cell differentiation, tumorigenesis, and apoptosis.[4] Polymorphisms in the expression of LCN2 are associated with many cancers such as breast, colorectal, pancreatic, ovarian, gastric, thyroid, ovarian, bladder and kidney cancers.[2]

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The pancreatic expression database: recent extensions and updates
Dayem Ullah AZ, et al.
Nursing New Zealand (Wellington, N.Z. : 1995) (2014)
Network based analyses of gene expression profile of LCN2 overexpression in esophageal squamous cell carcinoma
Wu B, et al.
Scientific Reports (2014)
The role of neutrophil gelatinase-associated lipocalin in predicting acute kidney injury in patients undergoing off-pump coronary artery bypass graft: A pilot study.
Jain V et al
Annals of Cardiac Anaesthesia, 19(2), 225-230 (2016)
Epigenetic induction of epithelial to mesenchymal transition by LCN2 mediates metastasis and tumorigenesis, which is abrogated by NF-?B inhibitor BRM270 in a xenograft model of lung adenocarcinoma.
Mongre RK et al
International Journal of Oncology, 48(1), 84-98 (2016)
María Garrido-Arandia et al.
Allergy, 74(11), 2167-2180 (2019-05-17)
Despite all the efforts made up to now, the reasons that facilitate a protein becoming an allergen have not been elucidated yet. Alt a 1 protein is the major fungal allergen responsible for chronic asthma, but little is known about

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service