Skip to Content
Merck
All Photos(1)

Key Documents

SRP6408

Sigma-Aldrich

ErbB4/HER4 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

MGC138404, p180erbB4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 70.6 kDa
observed mol wt 95-110 kDa by SDS-PAGE (reducing) (Glu26 is the predicted N-terminal .)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ErbB4(2066)

General description

Receptor tyrosine-protein kinase erbB-4 (ErbB4), also known as Her4, is a single-pass type I transmembrane glycoprotein that is a member of the ErbB family of tyrosine kinase receptors. ErbB family members serve as receptors for the epidermal growth factor (EGF) family of growth factors.[1] ErbB4 is expressed in normal skeletal muscle, heart, pituitary, brain and several breast carcinomas.[2] ERBB4 contains multiple furin-like cysteine rich domains, a tyrosine kinase domain, a phosphatidylinositol-3 kinase binding site and a PDZ domain binding motif.[3] The gene encoding this protein is localized on human chromosome 2q34.[4]

Biochem/physiol Actions

Receptor tyrosine-protein kinase erbB-4 (ErbB4) binds to and is activated by neuregulins-2 and -3, heparin-binding EGF-like growth factor and β-cellulin. Ligand binding induces a variety of cellular responses including mitogenesis and differentiation. Multiple proteolytic events allow for the release of a cytoplasmic fragment and an extracellular fragment.[3] ErbB4 appears to play important roles in neuronal development, development of the heart and cancer. Mutations in this gene have been associated with cancer.[3] Other single-nucleotide polymorphisms and a risk haplotype have been linked to schizophrenia.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ERBB4 Expression in Ovarian Serous Carcinoma Resistant to
Platinum-Based Therapy
Ozlen Saglam
Cancer Control : Journal of the Moffitt Cancer Center, 24(1), 89-95 (2017)
The epidermal growth factor receptor family
L A Bazley and W J Gullick
Endocrine-Related Cancer, S17-S27 (2005)
Ligand-specifi'c activation of HER4/pl80erbB4, a fourth member of
the epidermal growth factor receptor family
Gregory D. Plowman
Proceedings of the National Academy of Sciences of the USA, 90, 1746-1750 (1993)
A genome-wide association study identifies a breast cancer risk variant in ERBB4 at 2q34: results from the Seoul Breast Cancer Study
A genome-wide association study identifies a breast cancer risk variant in ERBB4 at 2q34: results from the Seoul Breast Cancer Study
Hyung-cheol Kim
Breast Cancer Research, 14, R56-R56 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service