Skip to Content
Merck
All Photos(2)

Key Documents

SRP6404

Sigma-Aldrich

EGFR/ErbB1 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

ERBB, ERBB1, HER1, PIG61, mENA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 69.5 kDa
observed mol wt 110-115 kDa (DTT-reduced.Protein migrates due to glycosylation.)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... EGFR(1956)

General description

EGFR (epidermal growth factor receptor) belongs to the ErbB family of receptor tyrosine kinases.[1] It is also known as ErbB1/HER1 (human epidermal growth factor receptor). It is located on human chromosome 7p11.2.[2] EGFR has a large extracellular ligand-binding domain, a single transmembrane segment, an intracellular juxtamembrane segment, a tyrosine kinase domain and a carboxy-terminal tail.[1]
The epidermal growth factor receptor (EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. The epidermal growth factor receptor is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: EGFR (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). Mutations affecting EGFR expression or activity could result in cancer.
The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells. (provided by RefSeq)

Biochem/physiol Actions

EGFR (epidermal growth factor receptor) plays an important role in ligand binding, dimerization and autophosphorylation.[1] It also plays an important role in several procarcinogenic cancers like gastric cancer. EGFR controls various functions of epithelial cell. It modulates the actions in bacterial infection and is associated with the initiation of macrophage.[3]

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biochemical characterization of EGF receptor in a water-soluble membrane model system
Scharadin TM, et al.
PLoS ONE (2017)
EGFR regulates macrophage activation and function in bacterial infection
Hardbower DM, et al.
The Journal of Clinical Investigation, 126(9), 3296-3312 (2016)
Chalita Washington et al.
PLoS genetics, 16(6), e1008715-e1008715 (2020-06-20)
Dysregulation of the Ras oncogene in development causes developmental disorders, "Rasopathies," whereas mutational activation or amplification of Ras in differentiated tissues causes cancer. Rabex-5 (also called RabGEF1) inhibits Ras by promoting Ras mono- and di-ubiquitination. We report here that Rabex-5-mediated
A chromosomal region 7p11.2 transcript map: its development and application to the study of EGFR amplicons in glioblastoma.
Eley GD
Neuro-Oncology, 4(2), 86-94 (2002)

Questions

  1. Is this product an extracellular EGFR?

    1 answer
    1. According to Uniprot, it is an extracellular protein, spanning from amino acid 25 to 645.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service