Skip to Content
Merck
All Photos(1)

Documents

SRP6357

Sigma-Aldrich

CXCL17 human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

Chemokine (C-X-C motif) ligand 17, DMC, Dcip1, MGC138300, UNQ473, VCC-1, VCC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥85% (SDS-PAGE)

form

liquid

mol wt

13.7 kDa (119 aa, 22-119 aa + His Tag)

packaging

pkg of 50 μg

concentration

0.25 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CXCL17(284340)

General description

Chemokine (C-X-C motif) ligand 17, also known as CXCL17, is a small cytokine belonging to the CXC chemokine family. CXCL17 is a 119 amino acid secreted protein that plays a role in angiogenesis. CXCL17 is expressed in skeletal muscle, trachea, lung, intestine and stomach, and is upregulated in duodenal mucosa of patients with acute cholera, as well as breast tumors. CXCL17 is considered a housekeeping chemokine for the movement of immature dendritic cells and non-activated blood monocytes into tissues, and is thought to be involved in the innate immune response. Recombinant human CXCL17 protein, fused to His-tag at N-terminus, was expressed in E.coli.

Physical form

0.25 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.4 M Urea and 10% glycerol.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service