Skip to Content
Merck
All Photos(1)

Key Documents

SRP6313

Sigma-Aldrich

Alpha 2 Antiplasmin from human plasma

≥95% (SDS-PAGE), lyophilized

Synonym(s):

AAP, Alpha-2-plasmin inhibitor, PLI, SERPINF2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€181.00

€181.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
100 μG
€181.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€181.00


Usually ships in 2 weeks.

Request a Bulk Order

Product Name

Alpha 2 Antiplasmin from human plasma, ≥95% (SDS-PAGE)

biological source

human

Assay

≥95% (SDS-PAGE)

form

lyophilized

potency

≥5.0 I.U. per mg

mol wt

70 kDa

packaging

pkg of 100 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SERPINF2(5345)

General description

α-2 antiplasmin (AAP) is a member of the Serpin superfamily. Liver and kidney are major sites of its production and other tissues such as muscle, intestine, central nervous system, and placenta also express its mRNA at a moderate level.[1] The tissue expression pattern indicates that it is a key regulator of plasmin mediated proteolysis in these tissues.[1] The AAP gene is mapped to human chromosome 17p13 and codes for a glycoprotein of single chain containing 464 amino acid residues.[2][3]

Biochem/physiol Actions

α-2 antiplasmin (AAP) is the primary physiological inhibitor of the serine protease plasmin, which is responsible for the dissolution of fibrin clots.[1] In addition to plasmin, it is also an efficient inhibitor of trypsin and chymotrypsin. α-antiplasmin-deficiency is a rare coagulation disorder which allows unrestrained fibrinolytic activity.[4] Individuals with this condition may receive therapeutic A2AP prior to surgery to prevent postoperative hemorrhaging.
α-2 antiplasmin (AAP) is the primary physiological inhibitor of the serine protease plasmin, which is responsible for the dissolution of fibrin clots.[1] It inhibits the action of protease by the formation In addition to plasmin, it is also an efficient inhibitor of trypsin and chymotrypsin. α-antiplasmin-deficiency is a rare coagulation disorder which allows unrestrained fibrinolytic activity.[2] AAP is known to inhibit fibrinogenolysis by preventing free plasmin circulation.[2]

Physical form

Lyophilized from 20 mM Bis-Tris, pH 6.4, with 200 mM NaCl.

Reconstitution

In water or aqueous buffer

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noncovalent interaction of ?2-antiplasmin with fibrin (ogen): localization of ?2-antiplasmin-binding sites.
Tsurupa G, et al.
Biochemistry, 49(35), 7643-7651 (2010)
Genome-wide loss of heterozygosity and copy number alteration in esophageal squamous cell carcinoma using the Affymetrix GeneChip Mapping 10 K array.
Hu N
BMC Genomics, 7, 299-299 (2006)
Ekaterina Mindel et al.
Journal of neuroscience research, 99(3), 966-976 (2020-12-10)
Many coagulation factor proteases are increased in the brain during ischemic stroke. One of these proteases is plasmin. In this study we established a novel method for direct quantitative measurement of plasmin activity in male mouse brain slices using a
Open-heart surgery in a patient with heterozygous alpha 2-antiplasmin deficiency. Perioperative strategies in the first reported case.
Shahian DM and Levine JD
Chest, 97(6), 1488-1490 (1990)
The kidney is a major site of alpha(2)-antiplasmin production.
Menoud PA
The Journal of Clinical Investigation, 97(11), 2478-2484 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service