Skip to Content
Merck
All Photos(1)

Key Documents

SRP6309

Sigma-Aldrich

Proteinase 3 from human neutrophils

≥95% (SDS-PAGE)

Synonym(s):

AGP7, C-ANCA antigen, Leukocyte proteinase 3, Neutrophil proteinase 4, P29, Wegener autoantigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

Assay

≥95% (SDS-PAGE)

form

frozen liquid

mol wt

29 kDa

packaging

pkg of 25 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AGP7(5657)

General description

Proteinase 3 is one of the serine proteinases localized in azurophil granules of polymorphonuclear leukocytes. It has been identified as the autoantigen of Wegener granulomatosis, a systemic vasculitis complicated by melomonocyte proliferation. It exhibits proteolytic activity towards a range of substrates. It is physiologically inhibited by α1-antitrypsin. The protein has been described as having antibiotic and growth-promoting properties and causing emphysema when administered by tracheal insufflation. Proteinase 3 (PR-3) is also known as myeloblastin and p29b.

Physical form

Frozen in 50 mM MES, pH 4.5 with 700 mM NaCl.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jeppe Thorlacius-Ussing et al.
Journal of cancer research and clinical oncology, 145(2), 383-392 (2018-11-24)
Elastin is a signature protein of lungs. Increased elastin turnover driven by altered proteolytic activity is an important part of lung tumorigenesis. Elastin-derived fragments have been shown to be pro-tumorigenic, however, little is known regarding the biomarker potential of such
Luis M Molinos-Albert et al.
Nature communications, 13(1), 1944-1944 (2022-04-13)
HIV-1 post-treatment controllers are rare individuals controlling HIV-1 infection for years after antiretroviral therapy interruption. Identification of immune correlates of control in post-treatment controllers could aid in designing effective HIV-1 vaccine and remission strategies. Here, we perform comprehensive immunoprofiling of

Questions

  1. Could you please provide the volume and concentration of the product with the code SRP6309-25ug?

    1 answer
    1. The volume and concentration are lot specific.

      Examples below:
      Lot: 6J30L72900
      Volume: 23.6 µL
      Concentration: 1.06 mg/mL

      Lot: 8A10L72900
      Volume: 22.9 µL
      Concentration: 1.0917 mg/mL

      The concentration of the newer lots should be on the vial label. The volume can then be calculated using the lot specific concentration and the pack size, which is 25 µg.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service