Skip to Content
Merck
All Photos(1)

Documents

SRP6092

Sigma-Aldrich

FKBP1B human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

FKBP12.6, FKBP1L, OTK4, PKBP1IL, Peptidyl prolyl cis-trans isomerase FKBP1B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

liquid

mol wt

14.2 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FKBP1B(2281)

General description

Peptidyl-prolyl cis-trans isomerase FKBP1B (FKBP1B) belongs to the immunophilin protein family which has a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP1B is a cis-trans prolyl isomerase which binds the immunosuppressants FK506 (tacrolimus) and rapamycin (sirolimus). FKBP1B is extremely similar to the FK506-binding protein 1A. The physiological role of FKBP1B is thought to be in excitation-contraction coupling in cardiac muscle.

Physical form

0.25 mg/mL solution in 20 mM Tris-HCl buffer (pH 8.0) containing 1 mM DTT and 20% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MGSSHHHHHH SSGLVPRGSH RSMGVEIETI SPGDGRTFPK KGQTCVVHYT GMLQNGKKFD SSRDRNKPFK FRIGKQEVIK GFEEGAAQMS LGQRAKLTCT PDVAYGATGH PGVIPPNATL IFDVELLNLE

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service