Skip to Content
Merck
All Photos(1)

Key Documents

SRP6002

Sigma-Aldrich

Chemerin human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

HP10433, RARRES2, TIG2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

16.0 kDa

packaging

pkg of 25 μg

impurities

Endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RARRES2(5919)

General description

Chemerin, which is also known as retinoic acid receptor responder 2 (RARRES2), is a chemoattractant expressed in white adipose, liver and lung tissues. It is an immunomodulating factor. The protein is a ligand for the G-protein coupled receptor known as ChemR23 (or chemokine-like receptor-1), which is expressed mainly on dendritic cells, macrophages and some adipocytes. The gene encoding it is localized on human chromosome 7q36.1.[1][2] Recombinant human Chemerin, produced in Escherichia.coli, is a non-glycosylated protein containing 138 amino acids and having a total molecular mass of 16kDa.

Biochem/physiol Actions

Chemerin activates chemerin-like receptor 1 (CMKLR1) and stimulates chemotaxis in macrophages, natural killer cells and immature dendritic cells. It is also involved in the activation of extracellular signal-regulated kinases, stimulation of blood vessel formation, migration and invasion.[1]

Physical form

Lyophilized from 0.2% TFA.

Reconstitution

Centrifuge the vial before opening. When reconstituting the product, gently pipet and wash down the sides of the vial to ensure full recovery of the protein into solution. It is recommended to reconstitute the lyophilized product with sterile H?O to a concentration of 0.1mg/ml, which can be further diluted into other aqueous solutions.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-xian Zhou et al.
Acta pharmacologica Sinica, 35(5), 653-663 (2014-03-25)
The chemerin receptor CMKLR1 is one type of G protein-coupled receptors abundant in monocyte-derived dendritic cells and macrophages, which plays a key role in the entry of a subset of immunodeficiency viruses including HIV/SIV into lymphocytes and macrophages. The aim
Jeremy Di Domizio et al.
Nature immunology, 21(9), 1034-1045 (2020-07-15)
Skin wounds heal by coordinated induction of inflammation and tissue repair, but the initiating events are poorly defined. Here we uncover a fundamental role of commensal skin microbiota in this process and show that it is mediated by the recruitment
Rui-Li Zhang et al.
Journal of cellular biochemistry, 120(12), 19621-19634 (2019-07-20)
Chemerin, a chemoattractant protein, is involved in endothelial dysfunction and vascular inflammation in pathological conditions. In a recent study, we observed the upregulation of chemerin in endothelial cells following in vitro treatment with Treponema pallidum. Here, we investigated the role
Association of Polymorphisms in STRA6 and RARRES2 Genes with Type 2 Diabetes in Southern Han Chinese.
Huang HW
BioMed Research International, 2016, 6589793-6589793 (2016)
Urszula Godlewska et al.
Frontiers in microbiology, 11, 1819-1819 (2020-08-28)
Chronic inflammatory skin diseases like psoriasis alter the local skin microbiome and lead to complications such as persistent infection with opportunistic/pathogenic bacteria. Disease-associated changes in microbiota may be due to downregulation of epidermal antimicrobial proteins/peptides, such as antimicrobial protein chemerin.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service