Skip to Content
Merck
All Photos(1)

Key Documents

SRP5366

Sigma-Aldrich

PSKH1, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€340.00

€340.00


Usually ships in 2 weeks.


Select a Size

Change View
20 μG
€340.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€340.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~72 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PSKH1(5681)

General description

PSKH1 is a member of the family of protein serine kinase H1 which was originally cloned by homology probing. PSKH1 has been localized to the endoplasmic reticulum (ER), the Golgi apparatus, and the plasma membrane (PM). PSKH1 has been shown to play a structural and regulatory role in maintenance of the Golgi apparatus, which is a key organelle within the secretory pathway. PSKH1 is in a cluster of 5 unrelated genes on 16q22.1 chromosome which is transcribed in the opposite direction from that of LCAT which is a member of the same cluster.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service