Skip to Content
Merck
All Photos(2)

Key Documents

SRP5333

Sigma-Aldrich

PTPRE, active, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp313F1310, FLJ57799, FLJ58245, HPTPE, P-PTP-epsilon, PTPE, R-PTP-EPSILON, RP11-380J17.1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€358.00

€358.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€358.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€358.00


Usually ships in 2 weeks.

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

6880-10320 nmol/min·mg

mol wt

~91 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPRE(5791)

General description

PTPRE or protein tyrosine phosphatase receptor type E is a member of the protein tyrosine phosphatase (PTP) family. This family of proteins are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. PTPRE is highly expressed in brain and testis. The regulatory roles of this PTP in Ras related signal transduction pathways, cytokines induced signaling, as well as the activation of voltage-gated K+ channels have been identified. PTPRE has a transmembrane segment and a cytoplasmic region containing duplicated PTPase domains.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service