Skip to Content
Merck
All Photos(2)

Key Documents

SRP5328

Sigma-Aldrich

NEK5, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC75495

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€358.00

€358.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€358.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

€358.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

80-120 nmol/min·mg

mol wt

~84 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NEK5(341676)

General description

NEK5 or NIMA (never in mitosis gene a)-related kinase 5 is a serine/threonine kinase that is involved in cell cycle regulation. NEK5 has been shown to interact with NEK6 and NEK5 are required for the ibo1/nek6 ectopic outgrowth phenotype in epidermal cells. NEK6 interacts and co-localizes with NEK5 in a transient expression assay. The ibo1-3 mutation markedly increases the interaction between NEK6 and NEK5. NEK6 forms heterodimers with NEK5 to regulate cortical microtubule organization possibly through the phosphorylation of β-tubulins. The NEK5 gene is conserved in chimpanzee, cow, mouse, rat, and chicken.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service