Skip to Content
Merck
All Photos(2)

Key Documents

SRP5294

Sigma-Aldrich

MEKK6, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ASK2, MAP3K6, MAPKKK6

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€358.00

€358.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€358.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€358.00


Usually ships in 2 weeks.

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

8-12 nmol/min·mg

mol wt

~67 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAP3K6(9064)

General description

MEKK6 or MAP3K6 (mitogen-activated protein kinase kinase kinase 6) is a member of the serine/threonine protein kinase family. MEKK6 was identified by its interaction with MAP3K5/ASK, a protein kinase and an activator of c-Jun kinase (MAPK7/JNK) and MAPK14/p38 kinase. MEKK6 cascades relay signals from the cell surface to the nucleus which result in gene transcription. MEKK6 activates the JNK and p38 pathway and induces apoptosis when overexpressed in stably transfected cells. MEKK6 transcript is expressed in several tissues, with the highest expression in heart and skeletal muscle.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service