Skip to Content
Merck
All Photos(2)

Key Documents

SRP5292

Sigma-Aldrich

PIP5K1A, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

RP11-68I18.9

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€405.00

€405.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€405.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€405.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

947-1281 nmol/min·mg

mol wt

~86 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PIP5K1A(8394)

General description

PIP5K1A or phosphatidylinositol-4-phosphate 5-kinase 1 alpha phosphorylates phosphatidylinositol-4-phosphate to form phosphatidylinositol 4,5-bisphosphate. PIP5K1 is required for Wnt3a-induced LRP6 phosphorylation at ser1490 in mammalian cells. Wnt3a stimulates the formation of phosphatidylinositol 4,5-bisphosphates through ′frizzled′ and ′dishevelled′, the latter of which directly interacts with and activated PIP5KI. PIP5K1A-mediated PIP production is crucial for HIV-1 entry and the early steps of infection in permissive lymphocytes. Overexpression of wild-type PI4P5K1A increases HIV-1 Env-mediated PIP production and enhances viral replication in primary lymphocytes.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service