Skip to Content
Merck
All Photos(2)

Key Documents

SRP5268

Sigma-Aldrich

HDAC9, active, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp779K1053, HD7, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, KIAA0744, MITR

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€509.00

€509.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€509.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

€509.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

199-299 RLU/min·ng

mol wt

~77 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HDAC9(9734)

General description

HDAC9 or MIRT belongs to the histone deacetylase /acuc/apha family and is a component of the histone deacetylase complex. HDAC9 gene is orthologous to the Xenopus and mouse MITR genes (MEF2-interacting transcription repressor). HDAC9 represses MEF2 activity through recruitment of multicomponent corepressor complexes that include CtBP and HDACs. HDAC9 is a calcium-sensitive transcriptional repressor of MEF2. Binding of calmodulin to HDAC9 leads to its dissociation from MEF2, relieving MEF2 from the transcriptional repression. HDAC9 is a signal-responsive transcriptional repressor in mouse skeletal muscle that is downregulated upon denervation.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service