Skip to Content
Merck
All Photos(2)

Key Documents

SRP5266

Sigma-Aldrich

HDAC2, active, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

RPD3, YAF1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€509.00

€509.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€509.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€509.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

3553-5520 RLU/min·ng

mol wt

~92 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HDAC2(3066)

General description

HDAC2 or Histone deacetylase 2 belongs to the histone deacetylase family that acts via the formation of large multiprotein complexes, and is responsible for the deacetylation of lysine residues at the N-terminal regions of core histones (H2A, H2B, H3 and H4). HDAC2 forms transcriptional repressor complexes by associating with many different proteins and plays an important role in transcriptional regulation, cell cycle progression and developmental events. HDAC2 functions in modulating synaptic plasticity and long-lasting changes of neural circuits, which in turn negatively regulates learning and memory. HDAC1 and HDAC2 are functionally redundant in cardiac growth and development and they maintain cardiomyocyte identity and function.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service