Skip to Content
Merck
All Photos(2)

Key Documents

SRP5247

Sigma-Aldrich

PDHK1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PDK1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€401.00

€401.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€401.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€401.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

1.4-2 nmol/min·mg

mol wt

~70 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDK1(5163)

Biochem/physiol Actions

PDHK1 or Pyruvate Dehydrogenase Kinase 1 is a member of the PDHK family that phosphorylate and inactivate the Pyruvate Dehydrogenase (PDH). PDH is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. PDHK1 is an important regulator of PDH in clonal beta-cells and the activity of PDHK1 and PDH are important for efficient metabolic coupling. Maintaining low PDHK1 expression/activity and keeping PDH in a dephosphorylated and active state, is important for beta-cells to achieve the metabolic flux rates necessary for maximal glucose-stimulated insulin secretion.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulrika Krus et al.
The Biochemical journal, 429(1), 205-213 (2010-04-27)
Tight coupling between cytosolic and mitochondrial metabolism is key for GSIS (glucose-stimulated insulin secretion). In the present study we examined the regulatory contribution of PDH (pyruvate dehydrogenase) kinase 1, a negative regulator of PDH, to metabolic coupling in 832/13 clonal
R Gudi et al.
The Journal of biological chemistry, 270(48), 28989-28994 (1995-12-01)
Recent evidence from this laboratory indicates that at least two isoenzymic forms of pyruvate dehydrogenase kinase (PDK1 and PDK2) may be involved in the regulation of enzymatic activity of mammalian pyruvate dehydrogenase complex by phosphorylation (Popov, K.M., Kedishvili, N.Y., Zhao

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service