Skip to Content
Merck
All Photos(2)

Key Documents

SRP5241

Sigma-Aldrich

CK1 γ 3, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CSNK1G3

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€354.00

€354.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€354.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€354.00


Usually ships in 2 weeks.

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

17-23 nmol/min·mg

mol wt

~73 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CSNK1G3(1456)

General description

CK1gamma3 is a member of the CK1 family of serine/threonine protein kinases which play an important role in diverse cell processes, including DNA replication and repair. CK1gamma3 is a ubiquitously expressed protein kinase found in the nuclei, cytoplasm and membrane fractions of eukaryotic cells. CK1gamma3 preferentially phosphorylates acidic substrates using ATP as a phosphate donor. The kinase domain of CKI isoforms have been shown to associate with protein kinase C-potentiated inhibitor protein of 17kDa, called CPI-17. CPI-17 specifically inhibits myosin light chain phosphatase and this effect is potentiated when it is phosphorylated on Thr-38 by protein kinases such as the CK1 isoforms.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cloning and chromosome mapping of the human casein kinase I gamma3 gene (CSNK1G3).
J Kusuda et al.
Cytogenetics and cell genetics, 83(1-2), 101-103 (1999-02-02)
Eva Zemlickova et al.
Biochemical and biophysical research communications, 316(1), 39-47 (2004-03-09)
The protein kinase C-potentiated inhibitor protein of 17kDa, called CPI-17, specifically inhibits myosin light chain phosphatase (MLCP). Phosphorylation of Thr-38 in vivo highly potentiates the ability of CPI-17 to inhibit MLCP. Thr-38 has been shown to be phosphorylated in vitro

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service