Skip to Content
Merck
All Photos(1)

Key Documents

SRP5213

Sigma-Aldrich

PHLPP2 (766-1043), GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

KIAA0931, PHLPPL

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€326.00

€326.00


Usually ships in 2 weeks.


Select a Size

Change View
50 μG
€326.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

€326.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~56 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PHLPP2(23035)

General description

PHLPP2 or PH domain and leucine rich repeat protein phosphatase 2 is the important regulators of Akt serine-threonine kinases and conventional/novel protein kinase C (PKC) isoforms. PHLPP1 and PHLPP2 differentially regulated AKT signaling by selectively dephosphorylating the hydrophobic motifs of AKT2 or AKT3. PHLPP may act as a tumor suppressor in several types of cancer due to its ability to block growth factor-induced signaling in cancer cells.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Brognard et al.
Molecular cell, 25(6), 917-931 (2007-03-28)
Akt/protein kinase B controls cell growth, proliferation, and survival. We recently discovered a novel phosphatase PHLPP, for PH domain leucine-rich repeat protein phosphatase, which terminates Akt signaling by directly dephosphorylating and inactivating Akt. Here we describe a second family member

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service