Skip to Content
Merck
All Photos(1)

Key Documents

SRP5188

Sigma-Aldrich

HSF1, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HSTF1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€268.00

€268.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
€268.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

€268.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~96 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HSF1(3297)

General description

HSF1 is a member of the heat shock transcription factor family. Protein-damaging stress lead to the activation of HSF1 which binds to upstream regulatory sequences in the promoters of heat shock genes leading to enhanced heat shock gene expression. The activation of HSF1 proceeds through a multi-step pathway, involving a monomer-to-trimer transition, nuclear accumulation and extensive posttranslational modifications. HSF1 activity is regulated at different levels by heat shock proteins and co-chaperones and is modulated further by a number of mechanisms involving other stress-regulated aspects of cell metabolism.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Voellmy
Handbook of experimental pharmacology, 172(172), 43-68 (2006-04-14)
The heat shock response is triggered primarily by nonnative proteins accumulating in a stressed cell and results in increased expression of heat shock proteins (Hsps), i.e., of chaperones capable of participating in the refolding or elimination of nonnative proteins. Best
L Pirkkala et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 15(7), 1118-1131 (2001-05-10)
The heat shock response, characterized by increased expression of heat shock proteins (Hsps) is induced by exposure of cells and tissues to extreme conditions that cause acute or chronic stress. Hsps function as molecular chaperones in regulating cellular homeostasis and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service