Skip to Content
Merck
All Photos(1)

Key Documents

SRP5133

Sigma-Aldrich

SMAD4, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DPC4, JIP, MADH4

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€236.00

€236.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
€236.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€236.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~95 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SMAD4(4089)

General description

SMAD4 is a member of the SMAD family and mediates signaling by the transforming growth factor-beta (TGFβ) superfamily and related ligands. TGFβ stimulation leads to phosphorylation and activation of SMAD1, SMAD2 and SMAD3, which form complexes with SMAD4 that accumulate in the nucleus and regulate transcription of target genes. SMAD signaling is negatively regulated by inhibitory SMADs and ubiquitin-mediated processes and proteasomal degradation of SMADs depend on the direct interaction of specific E3 ligases with SMADs. SMAD4 is targeted for degradation by multiple ubiquitin ligases that can simultaneously act on R-SMADs and signaling receptors. Such mechanisms of down-regulation of TGFβ signaling via degradation of SMADs may be critical for proper physiological response to this pathway.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Attisano et al.
Current opinion in cell biology, 10(2), 188-194 (1998-04-30)
The discovery of the Mothers against dpp (Mad) gene in Drosophila has opened a window on an entirely unique signalling pathway that functions to mediate responses to the tumour growth factor beta (TGF beta) superfamily. This pathway, which is comprised
C H Heldin et al.
Nature, 390(6659), 465-471 (1997-12-11)
The recent identification of the SMAD family of signal transducer proteins has unravelled the mechanisms by which transforming growth factor-beta (TGF-beta) signals from the cell membrane to the nucleus. Pathway-restricted SMADs are phosphorylated by specific cell-surface receptors that have serine/threonine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service