Skip to Content
Merck
All Photos(1)

Key Documents

SRP5129

Sigma-Aldrich

RhoD, His tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

RHOHP1, RHOM, Rho

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€177.00

€177.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
€177.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€177.00


Usually ships in 2 weeks.

Request a Bulk Order

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~22 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RHOD(29984)

General description

RhoD is a member of the Rho gene family that interacts with protein kinases and serves as a target of activated GTPase. Rho binds GTP and plays a critical role in muscle differentiation by regulating the expression of myogenin and MEF2 genes. Furthermore, Rho is involved in endosome dynamics and reorganization of the actin cytoskeleton and plays a role in coordinating membrane transport. mDia and ROCK are downstream effectors of Rho mediating Rho action on the actin cytoskeleton; mDia produces actin filaments by nucleation and polymerization and ROCK activate myosin to cross-link them for induction of actomyosin bundles and contractility.

Physical form

Supplied in 50mM MOPS, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuh Narumiya et al.
Cancer metastasis reviews, 28(1-2), 65-76 (2009-01-23)
The Rho subgroup of the Rho GTPases consisting of RhoA, RhoB and RhoC induces a specific type of actin cytoskeleton and carry out a variety of functions in the cell. mDia and ROCK are downstream effectors of Rho mediating Rho
Stéphane Gasman et al.
Nature cell biology, 5(3), 195-204 (2003-02-11)
Early endosomes move bidirectionally between the cell periphery and the interior through a mechanism regulated by the low molecular weight GTPase RhoD. Here, we identify a novel splice variant of human Diaphanous, hDia2C, which specifically binds to RhoD and is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service