Skip to Content
Merck
All Photos(1)

Key Documents

SRP5109

Sigma-Aldrich

p27KIP1, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CDKN1B, CDKN4, MEN1B

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€326.00

€326.00


Usually ships in 2 weeks.


Select a Size

Change View
50 μG
€326.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

€326.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~52 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CDKN1B(1027)

General description

KIP1 (cyclin-dependent kinase inhibitor 1B) is a kinesin-related motor protein required for mitotic spindle assembly and chromosome segregation. Many tumorigenic processes modulate cell-cycle progression by regulating the levels of the cyclin-dependent kinase inhibitor KIP1. KIP1 binds to and inhibits cyclinE-Cdk2 complex, cyclinA-CDK2 and cyclinD1-CDK4. The phosphorylation- and ubiquitination-dependent proteolysis of KIP1 is implicated in control of the G1-S transition in the cell cycle. KIP1 is critical for retinoblastoma protein (Rb)-induced cellular proliferative senescence.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Hoyt et al.
The Journal of cell biology, 118(1), 109-120 (1992-07-01)
Two Saccharomyces cerevisiae genes, CIN8 and KIP1 (a.k.a. CIN9), were identified by their requirement for normal chromosome segregation. Both genes encode polypeptides related to the heavy chain of the microtubule-based force-generating enzyme kinesin. Cin8p was found to be required for
A C Carrano et al.
Nature cell biology, 1(4), 193-199 (1999-11-13)
Degradation of the mammalian cyclin-dependent kinase (CDK) inhibitor p27 is required for the cellular transition from quiescence to the proliferative state. The ubiquitination and subsequent degradation of p27 depend on its phosphorylation by cyclin-CDK complexes. However, the ubiquitin-protein ligase necessary

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service