Skip to Content
Merck
All Photos(2)

Key Documents

SRP5074

Sigma-Aldrich

PTPN13 (2169-2485), active, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp686J1497, PNP1, PTP-BAS, PTP-BL, PTP1E, PTPL1, PTPLE

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€354.00

€354.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€354.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

€354.00


Usually ships in 2 weeks.

Request a Bulk Order

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

2040-2760 nmol/min·mg

mol wt

~61 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPN13(5783)

General description

PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. PTPN13 is a large protein that possesses a PTP domain at C-terminus, and multiple noncatalytic domains, which include a domain with similarity to band 4.1 superfamily of cytoskeletal-associated proteins, a region consisting of five PDZ domains, and a leucine zipper motif. PTPN13 binds to a negative regulatory domain in Fas that inhibits Fas-induced apoptosis.

Physical form

Supplied in 20mM MOPS, pH 7.5, 50mM NaCl, 10mM glutathione, 0.25mM DTT, 0.1mM PMSF, 30% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Inazawa et al.
Genomics, 31(2), 240-242 (1996-01-15)
PTPN13 is a protein tyrosine phosphatase that associates with the C-terminal negative regulatory domain in the Fas (APO-1/CD95) receptor. The PTPN13 protein contains six GLGF repeats that have been found in the rat postsynaptic density protein (PSD-95) and the Drosophila
J Saras et al.
The Journal of biological chemistry, 269(39), 24082-24089 (1994-09-30)
A novel cytoplasmic protein tyrosine phosphatase (PTP), PTPL1, was identified and cloned using a polymerase chain reaction-based approach. Overlapping cDNA clones encompass an open reading frame of 7398 base pairs, predicting a protein of 2466 amino acid residues with a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service