Skip to Content
Merck
All Photos(2)

Key Documents

SRP5042

Sigma-Aldrich

JNK2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

JNK-55, JNK2α, JNK2β, JNK2A, JNK2B, MAPK9, PRKM9, SAPK, p54a, p54aSAPK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

11-15 nmol/min·mg

mol wt

~70 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAPK9(5601)

Biochem/physiol Actions

JNK2 is a member of the JNK family that acts as an integration point for multiple biochemical signals, and is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK2 targets specific transcription factors, and mediates immediate-early gene expression in response to various cell stimuli. Both UV radiation and the proinflammatory cytokine TNFα can induce JNK2. JNK2 play a main role in the regulation of regional specific apoptosis during early brain development and providing protection against autoimmune diabetes.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Y Kuan et al.
Neuron, 22(4), 667-676 (1999-05-07)
The c-Jun NH2-terminal kinase (Jnk) family is implicated in apoptosis, but its function in brain development is unclear. Here, we address this issue using mutant mice lacking different members of the family (Jnk1, Jnk2, and Jnk3). Mice deficient in Jnk1
Anja Jaeschke et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(19), 6931-6935 (2005-05-04)
The c-Jun NH(2)-terminal kinase isoform (JNK) 1 is implicated in type 2 diabetes. However, a potential role for the JNK2 protein kinase in diabetes has not been established. Here, we demonstrate that JNK2 may play an important role in type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service