Skip to Content
Merck
All Photos(2)

Key Documents

SRP5036

Sigma-Aldrich

GRK7, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

GPRK7

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€354.00

€354.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€354.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€354.00


Usually ships in 2 weeks.

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

2.1-2.9 nmol/min·mg

mol wt

~89 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... GRK7(131890)

General description

GRK7 is a member of the G protein-coupled receptor kinase subfamily and is a Ser/Thr protein kinase. GRK7 is specifically expressed in the retina and has been shown to phosphorylate cone opsins and initiate their deactivation. GRK7 catalyzes rhodopsin phosphorylation in a light-dependent manner. GRK7 co-localizes with GRK1 in human cone outer segments. PKA phosphorylates GRK7 at Ser(23) and Ser(36) in vitro and phosphorylation of GRK7 by PKA reduces the ability of GRK7 to phosphorylate rhodopsin in vitro. Since exposure to light causes a decrease in cAMP levels in rod cells, it is proposed that phosphorylation of GRK7 by PKA occurs in the dark.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C K Chen et al.
Molecular vision, 7, 305-313 (2002-01-05)
Homozygous inactivation of the mouse gene for GRK1 (G protein-coupled receptor kinase 1, or rhodopsin kinase) causes severe defects in the recovery of cone phototransduction. However, electroretinographic (ERG) analyses of human oguchi patients with defective GRK1 alleles showed normal or
Thierry J Horner et al.
The Journal of biological chemistry, 280(31), 28241-28250 (2005-06-11)
Phosphorylation of G protein-coupled receptors is a critical step in the rapid termination of G protein signaling. In rod cells of the vertebrate retina, phosphorylation of rhodopsin is mediated by GRK1. In cone cells, either GRK1, GRK7, or both, depending

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service