Skip to Content
Merck
All Photos(2)

Key Documents

SRP5026

Sigma-Aldrich

EPHA1 (569-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

EPH, EPHT, EPHT1, MGC163163

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€354.00

€354.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€354.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€354.00


Usually ships in 2 weeks.

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

24-32 nmol/min·mg

mol wt

~72 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EPHA1(2041)

General description

EPHA1 is a member of the Eph family of receptor tyrosine kinases that have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the Eph subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. EPHA1 seems to be a marker of the differentiated normal epidermis and its downregulation in nonmelanoma skin cancer may contribute to carcinogenesis of these very frequent human tumors. EPHA1 represents a new potential prognostic marker and therapeutic target in nonmelanoma skin cancer.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanisms and functions of Eph and ephrin signalling.
Klas Kullander et al.
Nature reviews. Molecular cell biology, 3(7), 475-486 (2002-07-03)
Christian Hafner et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 19(10), 1369-1377 (2006-07-25)
Eph receptors and ephrin ligands represent the largest family of receptor tyrosine kinases. Beyond their well-defined meaning in developmental processes, these molecules also have important functions in adult human tissues and cancer. However, the Eph/ephrin expression profile in human skin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service