Skip to Content
Merck
All Photos(2)

Key Documents

SRP5012

Sigma-Aldrich

CDK9/CyclinK, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

C-2k, CCNK, CDC2L4, CPR4, MGC9113, PITALRE, TAK

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€401.00

€401.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€401.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€401.00


Usually ships in 2 weeks.

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

15-21 nmol/min·mg

mol wt

~67 kDa (CyclinK)
~68 kDa (CDK9)

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

CDK9/CyclinK, active, GST tagged human comprises recombinant full-length human CDK9 and cyclinK proteins co-expressedin Sf9 insect cells with an N-terminal GST tag. Cyclin-dependent protein kinase 9 (CDK9)/CyclinK is a member of the cyclin-dependent protein kinase (CDK) family. CDK9 is a component of the multiprotein complex mitogen-activated protein kinase 7 and transcription elongation factor β (TAK/P-TEFβ).[1]

Biochem/physiol Actions

CDKs are closely related to yeast CDk1, cdc28 and cdc2 and arean important regulators of the cell cycle.[2] CDK9 can modulate RNA polymerase II-directed transcription by phosphorylating the C-terminal domain of the largest subunit of RNA polymerase II.[3] CDK9 forms a complex with and is regulated by its regulatory subunit cyclin T or cyclin K. CDK9 also interacts with the human immune deficiency virus 1 (HIV-1) transactivating regulatory protein (Tat) protein, which suggests a possible involvement of this protein in AIDS.[1]

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ranadip Mandal et al.
Cellular and molecular life sciences : CMLS, 79(12), 597-597 (2022-11-19)
Cervical cancer is the fourth most frequently diagnosed and fatal gynecological cancer. 15-61% of all cases metastasize and develop chemoresistance, reducing the 5-year survival of cervical cancer patients to as low as 17%. Therefore, unraveling the mechanisms contributing to metastasis
The 7SK small nuclear RNA inhibits the CDK9/cyclin T1 kinase to control transcription
Yang Z, et al.
Nature, 414(6861), 317-322 (2001)
Phosphorylation of the RNA polymerase II carboxyl-terminal domain by CDK9 is directly responsible for human immunodeficiency virus type 1 Tat-activated transcriptional elongation
Kim YK, et al.
Molecular and Cellular Biology, 22(13), 4622-4637 (2002)
Z Yang et al.
Nature, 414(6861), 317-322 (2001-11-20)
The human positive transcription elongation factor P-TEFb, consisting of a CDK9/cyclin T1 heterodimer, functions as both a general and an HIV-1 Tat-specific transcription factor. P-TEFb activates transcription by phosphorylating RNA polymerase (Pol) II, leading to the formation of processive elongation
F Bullrich et al.
Cancer research, 55(6), 1199-1205 (1995-03-15)
Orderly progression through the cell cycle requires sequential activation and inactivation of cyclin-dependent kinases (cdks). This is achieved in part through the association of cdks with positive regulators called cyclins and inactivation of cyclin-cdk complexes by a rapidly growing number

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service