Skip to Content
Merck
All Photos(2)

Key Documents

SRP5010

Sigma-Aldrich

CDK2/CyclinA1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

p33(CDK2)

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€362.00

€362.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€362.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€362.00


Usually ships in 2 weeks.

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

70-95 nmol/min·mg

mol wt

~58 kDa (CDK1)
~81 kDa (CyclinA1)

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

CDK2 is a member of the Cyclin-Dependent Kinase family that is ubiquitously expressed. CDK2 is a catalytic subunit of the cyclin-dependent protein kinase complex, whose activity is restricted to the G1-S phase, and essential for cell cycle G1/S phase transition. CDK2 associates with and is regulated by the regulatory subunits of the complex including Cyclin A or E, CDK inhibitor p21Cip1 (CDKN1A) and p27Kip1 (CDKN1B). CDK2 phosphorylates multiple cellular substrates including SMAD3 and FOXO1. Phosphorylation of FOXO1 leads to its inhibition.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haojie Huang et al.
Science (New York, N.Y.), 314(5797), 294-297 (2006-10-14)
The function of cyclin-dependent kinase 2 (CDK2) is often abolished after DNA damage. The inhibition of CDK2 plays a central role in DNA damage-induced cell cycle arrest and DNA repair. However, whether CDK2 also influences the survival of cells under
B Levkau et al.
Molecular cell, 1(4), 553-563 (1998-07-14)
Apoptosis of human endothelial cells after growth factor deprivation is associated with rapid and dramatic up-regulation of cyclin A-associated cyclin-dependent kinase 2(cdk2) activity. In apoptotic cells, the C termini of the cdk inhibitors p21Cip1/Waf1 and p27Kip1 are truncated by specific

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service