SRP5001
AKT1, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
MGC99656, PKB, PRKBA, RAC, RAC-α
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
General description
v-akt murine thymoma viral oncogene homolog 1 (AKT1) is a Ser/Thr protein kinase, and is also called protein kinase B (PKB). This protein is activated when it is phosphorylated at serine 473 and threonine 308. This gene is localized on human chromosome 14q32.32.
Biochem/physiol Actions
v-akt murine thymoma viral oncogene homolog 1 (AKT1) is responsible for the phosphorylation and cytoplasmic localization of CKDN1A (cyclin-dependent kinase (CDK) inhibitor 1, also referred to as p21). However, p21 is one of the CDKNs which normally resides in the nucleus and its cytoplasmic localization, mediated by AKT1, is common to various cancers where it suppresses proteins essential for apoptosis. Studies show that up-regulation of AKT1 serves as a poor prognostic marker in breast cancer, which however, depends upon factors including population, hormone receptor status, and hormonal or trastuzumab therapy status.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gene expression profiling of mantle cell lymphoma cells reveals aberrant expression of genes from the PI3K-AKT, WNT and TGFbeta signalling pathways.
Rizzatti EG
British Journal of Haematology (2005)
The prognostic value of phosphorylated Akt in breast cancer: a systematic review.
Yang ZY
Scientific Reports (2015)
Wei Luo et al.
Nature immunology, 20(6), 736-746 (2019-04-24)
B cell antigen receptor (BCR) and CD40 signaling are rewired in germinal center (GC) B cells (GCBCs) to optimize selection for high-affinity B cells. In GCBC, BCR signals are constrained, but the mechanisms are not well understood. Here we describe
P J Coffer et al.
The Biochemical journal, 335 ( Pt 1), 1-13 (1998-09-22)
While a plethora of extracellular molecules exist that modulate cellular functions via binding to membrane receptors inside the cell, their actions are mediated by relatively few signalling mechanisms. One of these is activation of phosphatidylinositol 3-kinase (PI-3K), which results in
K E Anderson et al.
Current biology : CB, 8(12), 684-691 (1998-06-25)
Protein kinase B (PKB) is involved in the regulation of apoptosis, protein synthesis and glycogen metabolism in mammalian cells. Phosphoinositide-dependent protein kinase (PDK-1) activates PKB in a manner dependent on phosphatidylinositol 3,4,5-trisphosphate (PtdIns(3,4,5)P3), which is also needed for the translocation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service