Skip to Content
Merck
All Photos(1)

Key Documents

SRP4901

Sigma-Aldrich

Adiponectin human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

ACDC, APM-1, Acrp30, AdipoQ, GBP-28

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

lyophilized

mol wt

~30 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ADIPOQ(9370)

General description

Adiponectin (also called Acrp30, AdipoQ) is an adipocyte specific secreted protein that circulates in the plasma. It is induced during adipocyte differentiation and its secretion is stimulated by insulin.[1][2] The protein has a carboxyl-terminal globular domain and an amino-terminal collagen domain.[3] The gene is mapped to human chromosome 3q27.[4] Human adiponectin shares about 83% amino acid identity with that of mouse and about 90% with that of rat.

Application

Adiponectin human has been used to study the effect of adiponectin on androstenedione secretion and oxidative stress in rodent model.[5]

Biochem/physiol Actions

Adiponectin (also called Acrp30, AdipoQ) has antiatherosclerotic and anti-inflammatory properties.[6] It plays a role in various physiological processes such as energy homeostasis and obesity. Plasma levels of adiponectin are reduced in obese humans, and decreased levels are associated with insulin resistance and hyperinsulinemia.[2] It exhibits antidiabetic action by interacting with adiponectin receptors, AdipoR1 and AdipoR2.[3] Low levels of adiponectin are also linked with hypertension and dyslipidemia.[2] Mutation in the gene at position 276G is linked with risk for endometrial cancer.[7]

Physical form

Lyophilized from a 0.2 μm filtered solution in 25 mM Tris, 0.15 M NaCl, pH 7.4 containing 50 μg of bovine serum albumin (BSA) per 1 μg of adiponectin.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Recontitute in PBS containing at least 0.1% BSA to a concentration of 0.1-1.0 mg/mL. This stock solution can then be diluted into other aqueous buffers and stored at 4°C for 1 week or –20°C for future use.

Analysis Note

Activity of human recombinant adiponectin is measured by its ability to inhibit proliferation of mouse myeloid cell line M1. The ED50 for the effect is typically 1-7 μg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weena J Y Chen et al.
Cardiovascular diabetology, 10, 67-67 (2011-07-21)
Osteoprotegerin (OPG), a soluble member of the tumor necrosis factor receptor superfamily, is linked to cardiovascular disease. Negative associations exist between circulating OPG and cardiac function. The adipocytokine adiponectin (ADPN) is downregulated in type 2 diabetes mellitus (T2DM) and coronary
Heidi M Christensen et al.
BMC cardiovascular disorders, 13, 80-80 (2013-10-03)
Low body mass index (BMI) is associated with a poor outcome in chronic heart failure (CHF). An inverse association between BMI and adiponectin and N-terminal pro-B-type natriuretic peptide (NT-proBNP) has been reported. The aim of the present study was to
Plasma concentrations of a novel, adipose-specific protein, adiponectin, in type 2 diabetic patients.
Hotta K
Arteriosclerosis, Thrombosis, and Vascular Biology, 20, 1595-1599 (2000)
Eric R Hugo et al.
Environmental health perspectives, 116(12), 1642-1647 (2008-12-17)
The incidence of obesity has risen dramatically over the last few decades. This epidemic may be affected by exposure to xenobiotic chemicals. Bisphenol A (BPA), an endocrine disruptor, is detectable at nanomolar levels in human serum worldwide. Adiponectin is an
Thorhallur I Halldorsson et al.
Environmental health perspectives, 120(5), 668-673 (2012-02-07)
Perfluoroalkyl acids are persistent compounds used in various industrial -applications. Of these compounds, perfluorooctanoate (PFOA) is currently detected in humans worldwide. A recent study on low-dose developmental exposure to PFOA in mice reported increased weight and elevated biomarkers of adiposity

Articles

Lipid Induced Insulin Resistance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service