Skip to Content
Merck
All Photos(1)

Documents

SRP4365

Sigma-Aldrich

VEGF165 from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

MGC70609, VEGF, VEGF-A, VPF, Vascular endothelial growth factor A, Vascular permeability factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized

mol wt

~38.4 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

VEGF165 (vascular endothelial growth factor 165), also known as VEGF-A, is a homodimeric glycoprotein and is expressed as multiple splice variants. Recombinant rat VEGF is a double, non-glycosylated, polypeptide containing 164 amino acids and having a molecular mass of 38.4kDa.

Application

VEGF165 (vascular endothelial growth factor 165) has been used to study the effects of VEGF on the morphology and survival of neurons and glial cells.

Biochem/physiol Actions

VEGF165 (vascular endothelial growth factor 165) has been shown to have pro-angiogenic properties. It has a role in the proliferation of endothelial cells and neuroprotection.

Physical form

Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. The solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Different angiogenic pathways characterize superficial and invasive bladder cancer.
O'Brien T
Cancer Research, 55(3), 510-513 (1999)
Hongxiu Han et al.
The American journal of pathology, 191(3), 515-526 (2020-12-22)
Nerve infiltration into the tumor is a common feature of the tumor microenvironment. The mechanisms of axonogenesis in breast cancer remain unclear. We hypothesized that vascular endothelial growth factor (VEGF), as well as nerve growth factor (NGF), is involved in
Sarah A Figley et al.
PloS one, 9(5), e96137-e96137 (2014-05-23)
Following spinal cord injury (SCI) there are drastic changes that occur in the spinal microvasculature, including ischemia, hemorrhage, endothelial cell death and blood-spinal cord barrier disruption. Vascular endothelial growth factor-A (VEGF-A) is a pleiotropic factor recognized for its pro-angiogenic properties;
Larisa Antonova et al.
Pharmaceuticals (Basel, Switzerland), 14(4) (2021-04-04)
Tissue-engineered vascular grafts are widely tested as a promising substitute for both arterial bypass and replacement surgery. We previously demonstrated that incorporation of VEGF into electrospun tubular scaffolds from poly(3-hydroxybutyrate-co-3-hydroxyvalerate)/poly(ε-caprolactone) enhances formation of an endothelial cell monolayer. However, an overdose
Pauline Latzer et al.
CNS neuroscience & therapeutics, 22(6), 440-450 (2016-02-11)
Vascular endothelial growth factor (VEGF) is a hallmark of glioblastoma multiforme (GBM) and plays an important role in brain development and function. Recently, it has been reported that treatment of GBM patients with bevacizumab, an anti-VEGF antibody, may cause a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service