Skip to Content
Merck
All Photos(1)

Key Documents

SRP4363

Sigma-Aldrich

VEGF165 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

Folliculostellate cell-derived growth factor, Glioma-derived endothelial cell mitogen, VPF, Vascular Endothelial Growth Factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

1.0-7.0 ng/mL

mol wt

~38.2 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VEGFA(7422)

General description

The gene VEGFA (vascular endothelial growth factor A) is a member of VEGF family of growth factors. The gene is localized to human chromosome 6p21.1. It contains eight exons interspaced by seven introns, spanning a length of 14 kb. The critical region of 2.36 kbp in human promoter lacks a consensus TATA box. However, it contains various consensus-binding sites for many transcriptional regulators.

Biochem/physiol Actions

The gene VEGFA (vascular endothelial growth factor A) encodes a secreted mitogen that is found to participate in physiological and pathological angiogenesis. It serves as a growth factor for endothelial cells and functions in the formation of new blood vessels during embryonic development and adulthood. It binds to two tyrosine kinase receptors VEGFR1 and VEGFR2 and functions in angiogenesis and maintenance of vasculature.

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

VEGFA is necessary for chondrocyte survival during bone development.
Zelzer E
Development, 131, 2161-2171 (2004)
Processing of VEGF-A by matrix metalloproteinases regulates bioavailability and vascular patterning in tumors.
Lee S
The Journal of Cell Biology, 169, 681-691 (2005)
Genetic variants on chromosome 6p21.1 and 6p22.3 are associated with type 2 diabetes risk: a case-control study in Han Chinese.
Lu F
Journal of Human Genetics, 57, 320-325 (2012)
Vivian Lehmann et al.
Biotechnology progress, 35(2), e2745-e2745 (2018-11-14)
The only cure available for Type 1 diabetes involves the transplantation of islets of Langerhans isolated from donor organs. However, success rates are relatively low. Disconnection from vasculature upon isolation and insufficient rate of revascularization upon transplantation are thought to
Vivian Lehmann et al.
Biotechnology progress, 35(2), e2745-e2745 (2018-11-14)
The only cure available for Type 1 diabetes involves the transplantation of islets of Langerhans isolated from donor organs. However, success rates are relatively low. Disconnection from vasculature upon isolation and insufficient rate of revascularization upon transplantation are thought to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service